Anti-ANK3 Mouse Monoclonal Antibody [clone: S106-20]
Lieferant: US Biological
167575
USBI167575EA 587 EUR
USBI167575
Anti-ANK3 Mouse Monoclonal Antibody [clone: S106-20]
Antikörper
Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: monoclonal
Clone: S106-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat