Anti-Copper Transporting ATPase1 Mouse Monoclonal Antibody [clone: S60-4]

Lieferant: US Biological
167581
USBI167581EA 587 EUR
USBI167581
Anti-Copper Transporting ATPase1 Mouse Monoclonal Antibody [clone: S60-4]
Antikörper
Type: Primary
Antigen: Copper Transporting ATPase1
Clonality: monoclonal
Clone: S60-4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR