Anti-SYT6 Mouse Monoclonal Antibody [clone: S270-47]

Lieferant: US Biological
167610
USBI167610EA 514 EUR
USBI167610
Anti-SYT6 Mouse Monoclonal Antibody [clone: S270-47]
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: SYT6 (synaptotagmin VI)
Clonality: monoclonal
Clone: S270-47
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR