Anti-Hydroxyguanosine 8 Mouse Monoclonal Antibody [clone: 14B24]

Lieferant: US Biological
168252
USBI168252EA 728 EUR
USBI168252
Anti-Hydroxyguanosine 8 Mouse Monoclonal Antibody [clone: 14B24]
Antikörper
Recommended dilution: ELISA: 1:20000; Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: Hydroxyguanosine 8
Clonality: monoclonal
Clone: 14B24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR