Anti-KLH Rabbit Polyclonal Antibody

Lieferant: US Biological
168320
USBI168320EA 1020 EUR
USBI168320
Anti-KLH Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR