Anti-KLH Rabbit Polyclonal Antibody Rhodamine

Lieferant: US Biological
168326
USBI168326EA 722 EUR
USBI168326
Anti-KLH Rabbit Polyclonal Antibody Rhodamine
Antikörper
Recommended dilution: Immunofluorescence: 1:500-1:2500

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR