Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: US Biological
168489
USBI168489EA 910 EUR
USBI168489
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:3000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: RRM2B (ribonucleotide reductase M2 B (TP53 inducible))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR