Anti-SPANX, pan Rabbit Polyclonal Antibody

Lieferant: US Biological
168519
USBI168519EA 942 EUR
USBI168519
Anti-SPANX, pan Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: SPANX, pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR