Anti-Human TOP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
337594
USBI337594EA 775 EUR
USBI337594
Anti-Human TOP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Human TOP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR