Anti-ABCA1 Mouse Monoclonal Antibody [clone: 10K91]
Lieferant: US Biological
A0004-02J
USBIA0004-02JEA 499 EUR
USBIA0004-02J
Anti-ABCA1 Mouse Monoclonal Antibody [clone: 10K91]
Antikörper
  • Recommended dilution: ELISA: 3-6μg/ml; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: ABCA1 (ATP-binding cassette, sub-family A (ABC1), member 1)
Clonality: monoclonal
Clone: 10K91
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen