Anti-ABL2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
A0007-35K-PE
USBIA0007-35K-PEEA 884 EUR
USBIA0007-35K-PE
Anti-ABL2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: ABL2 (c-abl oncogene 2, non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR