Anti-ACHE Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A0526-41B
USBIA0526-41BEA 626 EUR
USBIA0526-41B
Anti-ACHE Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ACHE (acetylcholinesterase (Yt blood group))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR