Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 10K92]

Lieferant: US Biological
A0725-02T
USBIA0725-02TEA 540 EUR
USBIA0725-02T
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 10K92]
Antikörper
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: monoclonal
Clone: 10K92
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR