Anti-ACTN Mouse Monoclonal Antibody [clone: 10K98]
Lieferant: US Biological
A0762-01
USBIA0762-01EA 540 EUR
USBIA0762-01
Anti-ACTN Mouse Monoclonal Antibody [clone: 10K98]
Antikörper
Recommended dilution: Western Blot: 3.5 ?g/ml; Immunochemistry: 3.5-7?g/ml
Type: Primary
Antigen: ACTN (actinin, alpha)
Clonality: monoclonal
Clone: 10K98
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat