Anti-ATF2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A0855-42Q-BIOT
USBIA0855-42Q-BIOTEA 884 EUR
USBIA0855-42Q-BIOT
Anti-ATF2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR