Anti-ACVR2B Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A0855-97-BIOT
USBIA0855-97-BIOTEA 884 EUR
USBIA0855-97-BIOT
Anti-ACVR2B Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ACVR2B (activin A receptor, type IIB)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR