Anti-AIB 1 Mouse Monoclonal Antibody [clone: 6D102]

Lieferant: US Biological
A0857-06A
USBIA0857-06AEA 599 EUR
USBIA0857-06A
Anti-AIB 1 Mouse Monoclonal Antibody [clone: 6D102]
Antikörper
Type: Primary
Antigen: AIB 1
Clonality: monoclonal
Clone: 6D102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR