Anti-AIB 1 Mouse Monoclonal Antibody [clone: 4i291]

Lieferant: US Biological
A0857-07
USBIA0857-07EA 650 EUR
USBIA0857-07
Anti-AIB 1 Mouse Monoclonal Antibody [clone: 4i291]
Antikörper
Recommended dilution: Western Blot: 1-5 mg/ml

Type: Primary
Antigen: AIB 1
Clonality: monoclonal
Clone: 4i291
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR