Anti-AP2A1 Mouse Monoclonal Antibody [clone: 10K99]
Lieferant: US Biological
A0859-90B
USBIA0859-90BEA 540 EUR
USBIA0859-90B
Anti-AP2A1 Mouse Monoclonal Antibody [clone: 10K99]
Antikörper
Recommended dilution: Western Blot: 2-4?g/ml
Type: Primary
Antigen: AP2A1 (adaptor-related protein complex 2, alpha 1 subunit)
Clonality: monoclonal
Clone: 10K99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat