Anti-Adenovirus, Nuclear Mesh Mouse Monoclonal Antibody [clone: 10B71]

Lieferant: US Biological
A0880-23
USBIA0880-23EA 549 EUR
USBIA0880-23
Anti-Adenovirus, Nuclear Mesh Mouse Monoclonal Antibody [clone: 10B71]
Antikörper
Recommended dilution: Immunofluorescence: 1:20-1:80

Type: Primary
Antigen: Adenovirus, Nuclear Mesh
Clonality: monoclonal
Clone: 10B71
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR