Anti-ADIPOR1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
A0886-75G-BIOT
USBIA0886-75G-BIOTEA 903 EUR
USBIA0886-75G-BIOT
Anti-ADIPOR1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ADIPOR1 (adiponectin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human