Anti-ADRA2B Rabbit Polyclonal Antibody

Lieferant: US Biological
A0906-65
USBIA0906-65EA 667 EUR
USBIA0906-65
Anti-ADRA2B Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: ADRA2B (adrenoceptor alpha 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR