Anti-ADM Rabbit Polyclonal Antibody

Lieferant: US Biological
A0909-39C
USBIA0909-39CEA 871 EUR
USBIA0909-39C
Anti-ADM Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:500

Type: Primary
Antigen: ADM (adrenomedullin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR