Anti-Adrenomedullin N-20, Pro Rabbit Polyclonal Antibody

Lieferant: US Biological
A0909-40R
USBIA0909-40REA 836 EUR
USBIA0909-40R
Anti-Adrenomedullin N-20, Pro Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Adrenomedullin N-20, Pro
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR