Anti-ADM2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A0909-42N1
USBIA0909-42N1EA 1160 EUR
USBIA0909-42N1
Anti-ADM2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ADM2 (adrenomedullin 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR