Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 8J206]
Lieferant: US Biological
A0924-01D
USBIA0924-01DEA 614 EUR
USBIA0924-01D
Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 8J206]
Antikörper
Recommended dilution: Immunochemistry: 1:50-1:100
Type: Primary
Antigen: Aflatoxin B1 (aflatoxin B1)
Clonality: monoclonal
Clone: 8J206
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen