Anti-Aflatoxin B1/M1 Rat Monoclonal Antibody [clone: 3H57]

Lieferant: US Biological
A0925-01E
USBIA0925-01EEA 310 EUR
USBIA0925-01E
Anti-Aflatoxin B1/M1 Rat Monoclonal Antibody [clone: 3H57]
Antikörper
Type: Primary
Antigen: Aflatoxin B1/M1 (aflatoxin B1/M1)
Clonality: monoclonal
Clone: 3H57
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR