Anti-Aflatoxin B1/2 Mouse Monoclonal Antibody [clone: 10B82]

Lieferant: US Biological
A0925-04
USBIA0925-04EA 690 EUR
USBIA0925-04
Anti-Aflatoxin B1/2 Mouse Monoclonal Antibody [clone: 10B82]
Antikörper
Type: Primary
Antigen: Aflatoxin B1/2 (aflatoxin B1/2)
Clonality: monoclonal
Clone: 10B82
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR