Anti-Agmatine Rabbit Polyclonal Antibody

Lieferant: US Biological
A1059-56D
USBIA1059-56DEA 739 EUR
USBIA1059-56D
Anti-Agmatine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Agmatine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR