Anti-AGRN Mouse Monoclonal Antibody [clone: 10B87]
Lieferant: US Biological
A1059-60J
USBIA1059-60JEA 1180 EUR
USBIA1059-60J
Anti-AGRN Mouse Monoclonal Antibody [clone: 10B87]
Antikörper
Type: Primary
Antigen: AGRN (agrin)
Clonality: monoclonal
Clone: 10B87
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat