Anti-AKIRIN1 Rabbit Polyclonal Antibody
Lieferant: US Biological
A1105-80
USBIA1105-80EA 647 EUR
USBIA1105-80
Anti-AKIRIN1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml
Type: Primary
Antigen: AKIRIN1 (akirin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat