Anti-AKIRIN1 Rabbit Polyclonal Antibody

Lieferant: US Biological
A1105-80D
USBIA1105-80DEA 753 EUR
USBIA1105-80D
Anti-AKIRIN1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: AKIRIN1 (akirin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR