Anti-AKT2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A1125-25Y-BIOT
USBIA1125-25Y-BIOTEA 884 EUR
USBIA1125-25Y-BIOT
Anti-AKT2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR