Anti-AKT2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
A1125-25Y-PE
USBIA1125-25Y-PEEA 884 EUR
USBIA1125-25Y-PE
Anti-AKT2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR