Anti-GALP Rabbit Polyclonal Antibody Rhodamine

Lieferant: US Biological
A1232-21B6
USBIA1232-21B6EA 984 EUR
USBIA1232-21B6
Anti-GALP Rabbit Polyclonal Antibody Rhodamine
Antikörper
Type: Primary
Antigen: GALP (galanin-like peptide)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR