Anti-GALP Rabbit Polyclonal Antibody Rhodamine
Lieferant: US Biological
A1232-21C5
USBIA1232-21C5EA 984 EUR
USBIA1232-21C5
Anti-GALP Rabbit Polyclonal Antibody Rhodamine
Antikörper
Type: Primary
Antigen: GALP (galanin-like peptide)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat