Anti-Albuterol Sheep Polyclonal Antibody

Lieferant: US Biological
A1328-10A
USBIA1328-10AEA 763 EUR
USBIA1328-10A
Anti-Albuterol Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 10 ?g/ml

Type: Primary
Antigen: Albuterol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR