Anti-ADH Goat Polyclonal Antibody

Lieferant: US Biological
A1330-07B
USBIA1330-07BEA 640 EUR
USBIA1330-07B
Anti-ADH Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: ADH (alcohol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR