Anti-ADH Rabbit Polyclonal Antibody
Lieferant: US Biological
A1330-16A
USBIA1330-16AEA 818 EUR
USBIA1330-16A
Anti-ADH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADH (alcohol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen