Anti-ADH Rabbit Polyclonal Antibody

Lieferant: US Biological
A1330-17A
USBIA1330-17AEA 818 EUR
USBIA1330-17A
Anti-ADH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADH (alcohol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR