Anti-ADH Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A1330-18B
USBIA1330-18BEA 626 EUR
USBIA1330-18B
Anti-ADH Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ADH (alcohol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR