Anti-ALDH1A1 Goat Polyclonal Antibody

Lieferant: US Biological
A1334-33G
USBIA1334-33GEA 809 EUR
USBIA1334-33G
Anti-ALDH1A1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0. 03-0.1 ?g/ml; ELISA: 1:16000

Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR