Anti-ALDH3A1 Rabbit Polyclonal Antibody
Lieferant: US Biological
A1334-33M1
USBIA1334-33M1EA 667 EUR
USBIA1334-33M1
Anti-ALDH3A1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: ALDH3A1 (aldehyde dehydrogenase 3 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse