Anti-AKR1C3 Goat Polyclonal Antibody

Lieferant: US Biological
A1334-70O
USBIA1334-70OEA 809 EUR
USBIA1334-70O
Anti-AKR1C3 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 01-0.1 ?g/ml

Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR