Anti-AKR1C3 Goat Polyclonal Antibody

Lieferant: US Biological
A1334-70P1
USBIA1334-70P1EA 633 EUR
USBIA1334-70P1
Anti-AKR1C3 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1:32000

Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR