Anti-ALOX5AP Rabbit Polyclonal Antibody

Lieferant: US Biological
A1359-76J
USBIA1359-76JEA 568 EUR
USBIA1359-76J
Anti-ALOX5AP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:3000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR