Anti-ALOX5AP Goat Polyclonal Antibody

Lieferant: US Biological
A1359-76K
USBIA1359-76KEA 633 EUR
USBIA1359-76K
Anti-ALOX5AP Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Western Blot: 1:128000

Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR