Anti-STRADB Rabbit Polyclonal Antibody

Lieferant: US Biological
A1363-10F
USBIA1363-10FEA 649 EUR
USBIA1363-10F
Anti-STRADB Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: STRADB (STE20-related kinase adaptor beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR