Anti-AMELX Mouse Monoclonal Antibody [clone: 3B5]

Lieferant: US Biological
A1372-27A
USBIA1372-27AEA 623 EUR
USBIA1372-27A
Anti-AMELX Mouse Monoclonal Antibody [clone: 3B5]
Antikörper
Type: Primary
Antigen: AMELX (amelogenin, X-linked)
Clonality: monoclonal
Clone: 3B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR