Anti-ACY3 Rabbit Polyclonal Antibody

Lieferant: US Biological
A1374-08H
USBIA1374-08HEA 649 EUR
USBIA1374-08H
Anti-ACY3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: ACY3 (aspartoacylase (aminocyclase) 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR